2-(4-Hydroxyphenylazo)benzoic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Hydroxyazobenzene-2'-carboxylic acid, 2-(4-Hydroxyphenylazo)benzoic acid, 4'-Hydroxyazobenzene-2-carboxylic acid, 2-((4-Hydroxyphenyl)diazenyl)benzoic acid, HABA

H5126-1G H5126-25G H5126-5G
SIALH5126-1GEA 17 EUR
SIALH5126-1G SIALH5126-25G SIALH5126-5G
2-(4-Hydroxyphenylazo)benzoic acid, Sigma-Aldrich®
2-(4-Hydroxyphenylazo)benzoic acid
Formula: C₁₃H₁₀N₂O₃
MDL Number: MFCD00002428
CAS Number: 1634-82-8
EINECS: 216-655-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR