4-Allylveratrole, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
4-Allylveratrole, Eugenol methyll ether, 4-Prop-2-enylveratrole, 4-Allyl-1,2-Dimethoxybenzene, 4-Allyl-1,2-dimethoxybenzene, Methyl eugenol, Eugenol methyl ether, Methyleugenol
M-Clarity™ quality level = MQ100
284424-100G
284424-5G
SIAL284424-100GEA
146
EUR
SIAL284424-100G
SIAL284424-5G
4-Allylveratrole, Sigma-Aldrich®
4-Allylveratrole
Formula:
C₁₁H₁₄O₂ Boiling Pt: 255 °C (1013 hPa) Melting Pt: –4 °C Density: 1.036 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008652 CAS Number: 93-15-2 EINECS: 202-223-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...