Potassium iodate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
438464-25G
438464-5G
SIAL438464-25GEA
259
EUR
SIAL438464-25G
SIAL438464-5G
Potassium iodate, Sigma-Aldrich®
Potassium iodate
Formula:
KlO₃ MW: 214 g/mol Melting Pt: 560 Density: 3.98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011406 CAS Number: 7758-05-6 EINECS: 231-831-9 UN: 1479 ADR: 5.1 II Merck Index: 13,07726 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...