Periodic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: ortho-Periodic acid, Orthoperiodic acid
P0430-100G P0430-25G
SIALP0430-100GEA 206 EUR
SIALP0430-100G SIALP0430-25G
Periodic acid, Sigma-Aldrich®
Periodic acid
Formula: H₅IO₆
MW: 227.94 g/mol
Melting Pt: 122
Density: 1.3875 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011344
CAS Number: 10450-60-9
EINECS: 233-937-0
UN: 3085
ADR: 5.1 I
Merck Index: 13,07248
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39e1882cdf619581f467ea2429265a700b5205d5&vid=27e789fa0f30c08d62d5a8d0afc3acd8f6469c12&prid=web&clid=SAPDAM