Silver(I) oxide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: di-Silver oxide, Disilver oxide, Silver oxide
M-Clarity™ quality level = MQ100
223638-10G 223638-50G
SIAL223638-10GEA 118 EUR
SIAL223638-10G SIAL223638-50G
Silver(I) oxide, Sigma-Aldrich®
Silver(I) oxide
Formula: Ag₂O
MW: 231.74 g/mol
Melting Pt: 800…830 °C
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003404
CAS Number: 20667-12-3
EINECS: 243-957-1
UN: 1479
ADR: 5.1 II
Merck Index: 12,08665