2-Methyl-2-pentenoic acid ≥99%
Supplier: Thermo Fisher Scientific
Danger
A16587.09 A16587.18 A16587.30
A16587.09EA 32.9 EUR
A16587.09 A16587.18 A16587.30
2-Methyl-2-pentenoic acid ≥99%
2-Methyl-2-pentenoic acid
Purity: 99% CAS No.: 3142-72-1 Molecular Formula: C6H10O2 Formula Weight: 114.15 Flash Point: 107°(225°F) Odour: Characteristic Boiling Point: 120°/20mm UN No.: 3265 MDL No.: MFCD00002655
Formula: C₆H₁₀O₂
Boiling Pt: 120 °C (20 mmHg)
Density: 0.978
Flash Pt: 107 °C (225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002655
CAS Number: 3142-72-1
EINECS: 221-552-0
UN: 3265
ADR: 8,III