1-Tetradecyne tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
L02573.06
L02573.14
L02573.06EA
57.4
EUR
L02573.06
L02573.14
1-Tetradecyne tech. 90%, Technical Grade
1-Tetradecyne
Purity: 90%
CAS No.: 765-10-6
Molecular Formula: CH3(CH2)11C≡CH
Formula Weight: 194.36
Refractive Index: 1.4410
Boiling Point: 132-134°/14mm
MDL No.: MFCD00015076
Formula:
C₁₄H₂₆ Boiling Pt: 132…134 °C (14 mmHg) Density: 0.790 Storage Temperature: Ambient |
MDL Number:
MFCD00015076 CAS Number: 765-10-6 EINECS: 212-139-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...