2,6-Dibromophenylboronic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

704407-5G
SIAL704407-5GEA 356 EUR
SIAL704407-5G
2,6-Dibromophenylboronic acid, Sigma-Aldrich®
2,6-Dibromophenylboronic acid
Formula: C₆H₅BBr₂O₂
MDL Number: MFCD11656174
CAS Number: 851756-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR