Myrcene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 7-Methyl-3-methyleneocta-1,6-diene
M-Clarity™ quality level = MQ200
M100005-100ML M100005-500ML M100005-5ML
SIALM100005-100MLEA 37.4 EUR
SIALM100005-100ML SIALM100005-500ML SIALM100005-5ML
Myrcene, Sigma-Aldrich®
Myrcene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00008908
CAS Number: 123-35-3
EINECS: 204-622-5