Myrcene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
7-Methyl-3-methyleneocta-1,6-diene
M-Clarity™ quality level = MQ300
W276200-1KG-K
W276200-20KG-K
W276200-8KG-K
SIALW276200-1KG-KEA
137
EUR
SIALW276200-1KG-K
SIALW276200-20KG-K
SIALW276200-8KG-K
Myrcene, Sigma-Aldrich®
Myrcene
Formula:
C₁₀H₁₆ |
MDL Number:
MFCD00008908 CAS Number: 123-35-3 EINECS: 204-622-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...