2,6-Dibromo-4-(trifluoromethyl)aniline ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
L10658.03
L10658.06
L10658.03EA
21.3
EUR
L10658.03
L10658.06
2,6-Dibromo-4-(trifluoromethyl)aniline ≥97%
2,6-Dibromo-4-(trifluoromethyl)aniline
Purity: 97%
CAS No.: 72678-19-4
Molecular Formula: C7H4Br2F3N
Formula Weight: 318.93
Melting Point: 36-40°
Boiling Point: 64-65°/0.1mm
UN No.: 2811
MDL No.: MFCD00068181
Formula:
C₇H₄Br₂F₃N Boiling Pt: 64…65 °C (0.1 mmHg) Melting Pt: 36…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068181 CAS Number: 72678-19-4 UN: 2811 ADR: 6.1,III |