4-Bromo-2-(trifluoromethyl)aniline
Supplier: Thermo Scientific
Synonyms:
4-Bromo-α,α,α-trifluoro-o-toluidine, 2-Amino-5-bromobenzotrifluoride, 2-Amino-5-bromo-α,α,α-trifluorotoluene, 2-Amino-5-bromo-α,α,α-trifluortoluene
BTB02701.1
BTB02701.10
BTB02701.50
MAYBBTB02701.1EA
0
EUR
MAYBBTB02701.1
MAYBBTB02701.10
MAYBBTB02701.50
4-Bromo-2-(trifluoromethyl)aniline
4-Bromo-2-(trifluoromethyl)aniline
Formula:
C₇H₅BrF₃N Boiling Pt: 60.7…62 °C (1 mmHg) Density: 1.712 g/cm³ (25 °C) |
MDL Number:
MFCD00064393 CAS Number: 445-02-3 EINECS: 207-150-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...