2-Tolualdehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms: o-Tolualdehyde, 2-Methylbenzaldehyde
M-Clarity™ quality level = MQ100
117552-25G 117552-100G
SIAL117552-25GEA 61 EUR
SIAL117552-25G SIAL117552-100G
2-Tolualdehyde, Sigma-Aldrich®
2-Tolualdehyde
Formula: C₈H₈O
Boiling Pt: 201 °C (1013 hPa)
Melting Pt: –35 °C
Density: 1.0324 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003338
CAS Number: 529-20-4
EINECS: 208-452-2
Merck Index: 12,09665