Benzylacetone ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
B22777.0B
B22777.30
B22777.0BEA
112
EUR
B22777.0B
B22777.30
Benzylacetone ≥98%
Benzylacetone
Purity: 98%
CAS No.: 2550-26-7
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 105°(221°F)
Refractive Index: 1.5122
Boiling Point: 235-237°
Beilstein Reference: 1907123
MDL No.: MFCD00008790
Formula:
(C₆H₅)CH₂CH₂C(O)CH₃ Boiling Pt: 235…237 °C Melting Pt: –13 °C Density: 0.988 Flash Pt: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008790 CAS Number: 2550-26-7 EINECS: 219-847-4 |