4-Isopropylbenzoic acid 98+%
Supplier: Thermo Fisher Scientific
Warning
412800050 412800250
ACRO412800050EA 45.2 EUR
ACRO412800050 ACRO412800250
4-Isopropylbenzoic acid 98+%
4-Isopropylbenzoic acid
Formula: C₁₀H₁₂O₂
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 116…120 °C
Density: 1,162 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002564
CAS Number: 536-66-3
EINECS: 208-642-5
Merck Index: 12,02684
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 116 to 120 °C
HPLC ≥98.0 %