3-Methyl-4-nitrobenzoic acid
Supplier: Thermo Scientific
Synonyms:
3-Methyl-4-nitrobenzoic acid, 4-Nitro-m-toluic acid
SB01931.1
SB01931.10
SB01931.50
MAYBSB01931.1EA
0
EUR
MAYBSB01931.1
MAYBSB01931.10
MAYBSB01931.50
3-Methyl-4-nitrobenzoic acid
4-Nitro-m-toluic acid
Formula:
C₈H₇NO₄ Boiling Pt: 356 °C (1013 hPa) Melting Pt: 212…214 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007168 CAS Number: 3113-71-1 EINECS: 221-479-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...