(3-Chloropropyl)benzene ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1-Chloro-3-phenylpropane, 3-Phenylpropyl chloride, 3-Phenylpropyl Chloride
B20445.06 B20445.14 B20445.22
B20445.06EA 32.1 EUR
B20445.06 B20445.14 B20445.22
(3-Chloropropyl)benzene ≥98%
(3-Chloropropyl)benzene
Purity: 98% CAS No.: 104-52-9 Molecular Formula: C9H11Cl Formula Weight: 154.64 Flash Point: 87°(189°F) Refractive Index: 1.5220 Boiling Point: 218-220° MDL No.: MFCD00001001
Formula: ClH₂CCH₂CH₂(C₆H₅)
MW: 154.64 g/mol
Boiling Pt: 218…220 °C
Melting Pt: –27 °C
Density: 1.05
Flash Pt: 87 °C (189 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001001
CAS Number: 104-52-9
EINECS: 203-210-2