3,4-Dichlorotoluene ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
1,2-Dichloro-4-methylbenzene
A10763.0B
A10763.30
A10763.0BEA
128
EUR
A10763.0B
A10763.30
3,4-Dichlorotoluene ≥98%
3,4-Dichlorotoluene
Purity: 98%
CAS No.: 95-75-0
Molecular Formula: C7H6Cl2
Formula Weight: 161.03
Flash Point: 85°(185°F)
Refractive Index: 1.5470
Melting Point: ca -15°
Boiling Point: 200-202°
Beilstein Reference: 1931687
UN No.: 3082
MDL No.: MFCD00000556
Formula:
C₇H₆Cl₂ MW: 161.03 g/mol Boiling Pt: 200…202 °C Melting Pt: ∼–15 °C Density: 1.251 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000556 CAS Number: 95-75-0 EINECS: 202-447-9 UN: 3082 ADR: 9 III |