α,2,4-Trichlorotoluene ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98%
CAS No.: 94-99-5
Molecular Formula: C7H5Cl3
Formula Weight: 195.48
Flash Point: 190°(374°F)
Refractive Index: 1.5760
Odour: Pungent
Melting Point: -3°
Boiling Point: 248°
Beilstein Reference: 387220
UN No.: 3265
MDL No.: MFCD00000895
Formula:
C₇H₅Cl₃ Boiling Pt: 248 °C Melting Pt: –3 °C Density: 1.408 Flash Pt: 190 °C (374 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000895 CAS Number: 94-99-5 EINECS: 202-381-0 UN: 3265 ADR: 8,II |