3-Chlorotoluene ≥98%

Supplier: Thermo Fisher Scientific
Warning

A16991.0E A16991.22 A16991.36
A16991.0EEA 536 EUR
A16991.0E A16991.22 A16991.36
3-Chlorotoluene ≥98%
3-Chlorotoluene

Purity: 98% CAS No.: 108-41-8 Molecular Formula: C7H7Cl Formula Weight: 126.59 Flash Point: 50°(122°F) Refractive Index: 1.5220 Melting Point: -48° Boiling Point: 160-162° Beilstein Reference: 1903632 Merck Reference: 14,2171 UN No.: 2238 MDL No.: MFCD00000595

Formula: C₇H₇Cl
Boiling Pt: 160…162 °C
Melting Pt: –48 °C
Density: 1.072
Flash Pt: 50 °C (122 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000595
CAS Number: 108-41-8
EINECS: 203-580-5
UN: 2238
ADR: 3,III
Merck Index: 14,2171

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR