4-Chlorotoluene ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
1-Chloro-4-methylbenzene
L04672.22
L04672.36
L04672.0B
L04672.22EA
33
EUR
L04672.22
L04672.36
L04672.0B
4-Chlorotoluene ≥98%
4-Chlorotoluene
Purity: 98%
CAS No.: 106-43-4
Molecular Formula: C7H7Cl
Formula Weight: 126.59
Flash Point: 51°(124°F)
Refractive Index: 1.5200
Odour: Aromatic
Melting Point: 7-9°
Boiling Point: 161-163°
Beilstein Reference: 1903635
Merck Reference: 14,2171
UN No.: 2238
MDL No.: MFCD00000631
Formula:
C₇H₇Cl Boiling Pt: 161…163 °C Melting Pt: 7…9 °C Density: 1.069 Flash Pt: 51 °C (124 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000631 CAS Number: 106-43-4 EINECS: 203-397-0 UN: 2238 ADR: 3,III Merck Index: 14,2171 |