α-Chloro-2-fluorotoulene

Supplier: Thermo Scientific
Danger

Synonyms: 2-Fluorobenzyl chloride, 1-(Chloromethyl)-2-fluorobenzene, α-Chloro-o-fluorotoulene

SB00679.250 SB00679.1
MAYBSB00679.250EA 0 EUR
MAYBSB00679.250 MAYBSB00679.1
α-Chloro-2-fluorotoulene
α-Chloro-2-fluorotoulene
Formula: C₇H₆ClF
Boiling Pt: 86 °C (1013 hPa)
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000892
CAS Number: 345-35-7
EINECS: 206-460-0
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR