2-Chloro-α,α,α-trifluorotoluene ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1-Chloro-2-(trifluoromethyl)benzene, 2-Chlorobenzotrifluoride
A10753.0E
A10753.22
A10753.36
A10753.0EEA
270
EUR
A10753.0E
A10753.22
A10753.36
2-Chloro-α,α,α-trifluorotoluene ≥99%
2-Chloro-α,α,α-trifluorotoluene
Purity: 99%
CAS No.: 88-16-4
Molecular Formula: C7H4ClF3
Formula Weight: 180.56
Flash Point: 58°(136°F)
Refractive Index: 1.4560
Melting Point: -6°
Boiling Point: 151-153°
Beilstein Reference: 510993
UN No.: 2234
MDL No.: MFCD00000561
Formula:
C₇H₄ClF₃ Boiling Pt: 151…153 °C Melting Pt: –6 °C Density: 1.369 Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000561 CAS Number: 88-16-4 EINECS: 201-805-1 UN: 2234 ADR: 3,III |