3-Anisoyl chloride ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
3-Methoxybenzoyl chloride, m-Anisoyl chloride
B23571.09
B23571.18
B23571.30
B23571.09EA
47.4
EUR
B23571.09
B23571.18
B23571.30
3-Anisoyl chloride ≥99%
3-Anisoyl chloride
Purity: 99%
CAS No.: 1711-05-3
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
Flash Point: 92°(197°F)
Refractive Index: 1.5586
Boiling Point: 123-125°/15mm
Beilstein Reference: 386659
UN No.: 1729
MDL No.: MFCD00000673
Formula:
C₈H₇ClO₂ MW: 170.6 g/mol Boiling Pt: 123…125 °C (15 mmHg) Density: 1.214 Flash Pt: 92 °C (197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000673 CAS Number: 1711-05-3 EINECS: 216-975-2 UN: 1729 ADR: 8 II |