2-Fluoro-5-nitrotoluene ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1-Fluoro-2-methyl-4-nitrobenzene
A14649.09
A14649.18
A14649.09EA
48.9
EUR
A14649.09
A14649.18
2-Fluoro-5-nitrotoluene ≥98%
2-Fluoro-5-nitrotoluene
Purity: 98+%
CAS No.: 455-88-9
Molecular Formula: C7H6FNO2
Formula Weight: 155.13
Flash Point: 105°(221°F)
Melting Point: 37-40°
Boiling Point: 98-101°/13mm
Beilstein Reference: 1940341
UN No.: 2811
MDL No.: MFCD00007284
Formula:
C₇H₆FNO₂ Boiling Pt: 98…101 °C (13 mmHg) Melting Pt: 37…40 °C Flash Pt: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007284 CAS Number: 455-88-9 EINECS: 207-251-7 UN: 2811 ADR: 6.1,III |