2-Fluoro-4-nitrotoluene ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A14464.14
A14464.06
A14464.22
A14464.14EA
76.2
EUR
A14464.14
A14464.06
A14464.22
2-Fluoro-4-nitrotoluene ≥98%
2-Fluoro-4-nitrotoluene
Purity: 98%
CAS No.: 1427-07-2
Molecular Formula: C7H6FNO2
Formula Weight: 155.13
Flash Point: 73°(163°F)
Melting Point: 32-37°
Boiling Point: 56-58°/1mm
Beilstein Reference: 2250156
UN No.: 2811
MDL No.: MFCD00007199
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol Boiling Pt: 56…58 °C (1 mmHg) Melting Pt: 32…37 °C Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007199 CAS Number: 1427-07-2 EINECS: 215-845-2 UN: 2811 ADR: 6.1 III |