Benzylamine 99%, pure
Supplier: Thermo Fisher Scientific
Danger
Synonyms: α-Aminotoluene
105851000 105855000 105850025 105850100
ACRO105851000EA 36.5 EUR
ACRO105851000 ACRO105855000 ACRO105850025 ACRO105850100
Benzylamine 99%, pure
Benzylamine
Formula: (C₆H₅)CH₂NH₂
MW: 107.16 g/mol
Boiling Pt: 182…185 °C (760 mmHg)
Melting Pt: –30 °C
Density: 0.98 g/cm³
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00008106
CAS Number: 100-46-9
EINECS: 202-854-1
UN: 2735
ADR: 8 II
Merck Index: 13,01127
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5420 to 1.5450 (20°C, 589 nm)
GC ≥98.5 %