tert-Butylbenzene ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 99%
CAS No.: 98-06-6
Molecular Formula: C10H14
Formula Weight: 134.22
Flash Point: 34°(93°F)
Refractive Index: 1.4920
Odour: Characteristic
Melting Point: -58°
Boiling Point: 168-169°
Beilstein Reference: 1421537
Merck Reference: 14,1551
UN No.: 2709
MDL No.: MFCD00008816
Formula:
C₁₀H₁₄ Boiling Pt: 168…169 °C Melting Pt: –58 °C Density: 0.867 Flash Pt: 34 °C (93 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008816 CAS Number: 98-06-6 EINECS: 202-632-4 UN: 2709 ADR: 3,III Merck Index: 14,1551 |