3,4-Dimethylbenzaldehyde ≥97%

Supplier: Thermo Fisher Scientific

A15388.06 A15388.14
A15388.06EA 89.3 EUR
A15388.06 A15388.14
3,4-Dimethylbenzaldehyde ≥97%
3,4-Dimethylbenzaldehyde

Purity: 97% CAS No.: 5973-71-7 Molecular Formula: C9H10O Formula Weight: 134.18 Refractive Index: 1.5510 Odour: Almond-like Boiling Point: 226° Beilstein Reference: 1634270 MDL No.: MFCD00016612

Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 226 °C
Density: 1.017
Storage Temperature: Ambient
MDL Number: MFCD00016612
CAS Number: 5973-71-7
EINECS: 227-770-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR