Trimethylaluminium 1.0 M in heptane, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Trimethylaluminum
189271000 189278000
189271000.EA 80.2 EUR
189271000. 189278000.
Trimethylaluminium 1.0 M in heptane, AcroSeal®
Trimethylaluminium
Formula: C₃H₉Al
MW: 72.09 g/mol
Boiling Pt: 125
Melting Pt: 15
Density: 0.688 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008252
CAS Number: 75-24-1
EINECS: 200-853-0
UN: 3399
ADR: 4.3 I
Order Now
Specification Test Results
Appearance Clear colorless Solution
Titration Complexometric 9.5 to 10.5 % w/w (on Al basis)
HPLC ≥97.5 %