Selenourea 98+%
Supplier: Thermo Fisher Scientific
Danger
213170010 213170050
ACRO213170010EA 57 EUR
ACRO213170010 ACRO213170050
Selenourea 98+%
Selenourea
Formula: CH₄N₂Se
Melting Pt: 214…215 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00008065
CAS Number: 630-10-4
EINECS: 211-129-9
UN: 3283
ADR: 6.1,II
Order Now
Specification Test Results
Appearance White to pink to grey Powder or crystals or needles
Infrared spectrum Conforms
Assay ≥98.0 %