Decafluoro-4,6-bis(trifluoromethyl)cyclohexane tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
|
Warning
|
45477.18
45477.30
45477.18EA
89
EUR
45477.18
45477.30
Decafluoro-4,6-bis(trifluoromethyl)cyclohexane tech. 90%, Technical Grade
Decafluoro-4,6-bis(trifluoromethyl)cyclohexane
Purity: 90%
CAS No.: 335-27-3
Molecular Formula: C8F16
Formula Weight: 400.06
Physical Form: Liquid
Density: 1.828
Melting Point: -55°
Boiling Point: 101-102°
Beilstein Reference: 2064147
MDL No.: MFCD00001469
Formula:
C₈F₁₆ Boiling Pt: 101…102 °C Melting Pt: –55 °C Density: 1.828 Storage Temperature: Ambient |
MDL Number:
MFCD00001469 CAS Number: 335-27-3 EINECS: 206-386-9 |