4-Aminomorpholine 95%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N-Aminomorpholine

B21381.06 B21381.14 B21381.22
B21381.06EA 73 EUR
B21381.06 B21381.14 B21381.22
4-Aminomorpholine 95%
4-Aminomorpholine

Purity: 98% CAS No.: 4319-49-7 Molecular Formula: C4H10N2O Formula Weight: 102.14 Storage Temperature: -30°C to -10°C Flash Point: 58°(137°F) Refractive Index: 1.4785 Boiling Point: 168° UN No.: 2733 MDL No.: MFCD00006174

Formula: C₄H₁₀N₂O
Boiling Pt: 168 °C
Density: 1.059
Flash Pt: 58 °C (137 °F)
MDL Number: MFCD00006174
CAS Number: 4319-49-7
EINECS: 224-347-4
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR