N-(3-Aminopropyl)morpholine 98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 3-(4-Morpholinyl)propylamine, 3-Morpholinopropan-1-amine, 3-Morpholino propylamine
B23485.18 B23485.30 AB131469.0050
B23485.18EA 74 EUR
B23485.18 B23485.30 ABCRAB131469.0050
N-(3-Aminopropyl)morpholine 98%
N-(3-Aminopropyl)morpholine
Purity: 98% CAS No.: 123-00-2 Molecular Formula: C7H16N2O Formula Weight: 144.22 Flash Point: 98°(210°F) Refractive Index: 1.4761 Melting Point: -15° Boiling Point: 223-225° UN No.: 2735 MDL No.: MFCD00006184
Formula: C₇H₁₆N₂O
MW: 144.22 g/mol
Boiling Pt: 223…225 °C
Melting Pt: –15 °C
Density: 0.987
Flash Pt: 98 °C (210 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006184
CAS Number: 123-00-2
EINECS: 204-590-2
ADR: 8 II