4-Nonyloxyaniline ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
L09126.09
L09126.04
L09126.09EA
245
EUR
L09126.09
L09126.04
4-Nonyloxyaniline ≥98%
4-Nonyloxyaniline
Purity: 98%
CAS No.: 50262-67-4
Molecular Formula: C15H25NO
Formula Weight: 235.37
Melting Point: 36-42°
Boiling Point: 176-179°/2mm
UN No.: 2811
MDL No.: MFCD00043622
Formula:
C₁₅H₂₅NO MW: 235.37 g/mol Boiling Pt: 176…179 °C (2 mmHg) Melting Pt: 36…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043622 CAS Number: 50262-67-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...