4-Morpholinoaniline

Supplier: Thermo Scientific

Synonyms: 4-(4-Morpholinyl)aniline

BTB01373.1 BTB01373EA BTB01373.50
MAYBBTB01373.1EA 0 EUR
MAYBBTB01373.1 MAYBBTB01373EA MAYBBTB01373.50
4-Morpholinoaniline
4-Morpholinoaniline
Formula: C₁₀H₁₄N₂O
MDL Number: MFCD00006169
CAS Number: 2524-67-6
EINECS: 219-760-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR