2,3-Dimethylbenzoic acid

Supplier: Thermo Scientific
Warning

TL00856.1 TL00856.10 TL00856.50
MAYBTL00856.1EA 0 EUR
MAYBTL00856.1 MAYBTL00856.10 MAYBTL00856.50
2,3-Dimethylbenzoic acid
2,3-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 282
Melting Pt: 143…147 °C
Density: 1.283 g/cm³ (20 °C)
MDL Number: MFCD00002479
CAS Number: 603-79-2
EINECS: 210-058-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR