5-Bromothiophene-2-carbaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms: 5-Bromo-2-thiophenecarboxaldehyde
M-Clarity™ quality level = MQ100
152625-100G 152625-25G
SIAL152625-100GEA 651 EUR
SIAL152625-100G SIAL152625-25G
5-Bromothiophene-2-carbaldehyde, Sigma-Aldrich®
5-Bromothiophene-2-carbaldehyde
Formula: C₅H₃BrOS
MDL Number: MFCD00005432
CAS Number: 4701-17-1
EINECS: 225-176-8