Trimethylaluminium 2 mol/l in toluene, Sigma-Aldrich®
Supplier: Merck
Synonyms: Trimethylaluminum
M-Clarity™ quality level = MQ200
198048-100ML 198048-1L 198048-800ML 198048-8L
SIAL198048-100MLEA 78.3 EUR
SIAL198048-100ML SIAL198048-1L SIAL198048-800ML SIAL198048-8L
Trimethylaluminium 2 mol/l in toluene, Sigma-Aldrich®
Trimethylaluminium
Formula: C₃H₉Al
MW: 72.09 g/mol
Boiling Pt: 125
Melting Pt: 15
Density: 0.748
MDL Number: MFCD00008252
CAS Number: 75-24-1
EINECS: 200-853-0
UN: 3399
ADR: 4.3 I