Trimethylaluminium 2 mol/l in hexane, Sigma-Aldrich®
Supplier: Merck
Synonyms: Trimethylaluminum
M-Clarity™ quality level = MQ100
268569-100ML 268569-800ML
SIAL268569-100MLEA 120 EUR
SIAL268569-100ML SIAL268569-800ML
Trimethylaluminium 2 mol/l in hexane, Sigma-Aldrich®
Trimethylaluminium
Formula: C₃H₉Al
MW: 72.09 g/mol
Boiling Pt: 125
Melting Pt: 15
Density: 0.748
MDL Number: MFCD00008252
CAS Number: 75-24-1
EINECS: 200-853-0
UN: 3399
ADR: 4.3 I