Isobutylnitrite stabilised, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

327190-100ML 327190-500ML
SIAL327190-100MLEA 57.6 EUR
SIAL327190-100ML SIAL327190-500ML
Isobutylnitrite stabilised, Sigma-Aldrich®
IsobutylNitrite
Formula: C₄H₉NO₂
MDL Number: MFCD00002056
CAS Number: 542-56-3
EINECS: 208-819-7
UN: 2351
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR