3-Bromophenethylamine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

534080-10G 534080-5G
SIAL534080-10GEA 295 EUR
SIAL534080-10G SIAL534080-5G
3-Bromophenethylamine, Sigma-Aldrich®
3-Bromophenethylamine
Formula: C₈H₁₀BrN
MDL Number: MFCD01310790
CAS Number: 58971-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR