4-Methylphenethyl bromide, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-(2-Bromoethyl)-4-methylbenzene

653810-1G 653810-5G
SIAL653810-1GEA 43.5 EUR
SIAL653810-1G SIAL653810-5G
4-Methylphenethyl bromide, Sigma-Aldrich®
4-Methylphenethyl bromide
Formula: C₉H₁₁Br
MDL Number: MFCD00050368
CAS Number: 6529-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR