DURAN® Flasks, Round Bottom, with Two Inclined Side Necks and Standard Ground Joints
Supplier: DWK Life Sciences
241882703 241883605 241883708 241884301 241885306 241885503
SCOT241882703EA 69.4 EUR
SCOT241882703 SCOT241883605 SCOT241883708 SCOT241884301 SCOT241885306 SCOT241885503
DURAN® Flasks, Round Bottom, with Two Inclined Side Necks and Standard Ground Joints
Flasks Round Bottom Flasks
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • USP Standard
With white, matt labelling area.