o-Xylene 99%, pure
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 1,2-Dimethylbenzene
140990010 140990025 140990100
ACRO140990010EA 59 EUR
ACRO140990010 ACRO140990025 ACRO140990100
o-Xylene 99%, pure
o-Xylene
Formula: C₈H₁₀
Boiling Pt: 143…145 °C (760 mmHg)
Melting Pt: –25 °C
Density: 0.878 g/cm³
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00008519
CAS Number: 95-47-6
EINECS: 202-422-2
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dd6c62d1c62b6e7f71df7e157aae77162b3bcf1&vid=3ec8c09bb94f634e9f90b6f9e8320cd626f276d0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5035 to 1.5065 (20°C, 589 nm)
GC ≥98.5 %
Residue after evaporation ≤0.01 %