1,4-Diethylbenzene ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A13343.06
A13343.14
A13343.22
A13343.06EA
74.5
EUR
A13343.06
A13343.14
A13343.22
1,4-Diethylbenzene ≥98%
1,4-Diethylbenzene
Purity: 98%
CAS No.: 105-05-5
Molecular Formula: C10H14
Formula Weight: 134.22
Flash Point: 55°(131°F)
Refractive Index: 1.4950
Odour: Aromatic
Melting Point: -43°
Boiling Point: 182-184°
Beilstein Reference: 1903396
UN No.: 2049
MDL No.: MFCD00009264
Formula:
C₁₀H₁₄ Boiling Pt: 182…184 °C Melting Pt: –43 °C Density: 0.862 Flash Pt: 55 °C (131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009264 CAS Number: 105-05-5 EINECS: 203-265-2 UN: 2049 ADR: 3,III |