1,2,3,4-Tetramethylbenzene 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
L19756.06
L19756.14
L19756.06EA
162
EUR
L19756.06
L19756.14
1,2,3,4-Tetramethylbenzene 95%
1,2,3,4-Tetramethylbenzene
Purity: 95%
CAS No.: 488-23-3
Molecular Formula: C10H14
Formula Weight: 134.22
Flash Point: 68°(154°F)
Refractive Index: 1.5190
Boiling Point: 203°
Beilstein Reference: 1904390
MDL No.: MFCD00008521
Formula:
C₁₀H₁₄ Boiling Pt: 203 °C Melting Pt: –6 °C Density: 0.900 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008521 CAS Number: 488-23-3 EINECS: 207-673-1 |