6-Methyl-5-nitroquinoline

Supplier: Thermo Scientific

BTB10313DA BTB10313EA
MAYBBTB10313DAEA 0 EUR
MAYBBTB10313DA MAYBBTB10313EA
6-Methyl-5-nitroquinoline
6-Methyl-5-nitroquinoline
Formula: C₁₀H₈N₂O₂
MDL Number: MFCD00024021
CAS Number: 23141-61-9
EINECS: 245-447-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR