Iodocyclohexane 98% stabilised
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Cyclohexyl iodide
218490250 218491000
ACRO218490250EA 69.3 EUR
ACRO218490250 ACRO218491000
Iodocyclohexane 98% stabilised
Iodocyclohexane
Formula: C₆H₁₁I
MW: 210.06 g/mol
Boiling Pt: 80…81 °C (20 mmHg)
Density: 1.62 g/cm³
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003826
CAS Number: 626-62-0
EINECS: 210-957-8
Order Now
Specification Test Results
Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5490 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer Copper