Cyclopropylamine ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Aminocyclopropane
A13844.30
A13844.09
A13844.18
A13844.30EA
487
EUR
A13844.30
A13844.09
A13844.18
Cyclopropylamine ≥98%
Cyclopropylamine
Purity: 98+%
CAS No.: 765-30-0
Molecular Formula: C3H7N
Formula Weight: 57.10
Flash Point: -25°(-13°F)
Refractive Index: 1.4206
Melting Point: -10°
Boiling Point: 49-50°
Beilstein Reference: 741858
UN No.: 2733
MDL No.: MFCD00001301
Formula:
C₃H₇N Boiling Pt: 49…50 °C Melting Pt: –10 °C Density: 0.824 Flash Pt: –25 °C (–13 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001301 CAS Number: 765-30-0 EINECS: 212-142-2 UN: 2733 ADR: 3,II |